Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MICAL3 SIRNA Assays ›
Gene Symbol:
MICAL3
Gene Name:
microtubule associated monooxygenase, calponin and LIM domain containing 3
Gene Aliases:
MICAL-3
Chromosome Location:
Chr.22: 17787649 - 18024559 on Build GRCh38
UniGene ID:
Hs.528024
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s61182
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MICAL3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_015241.2
NP_056056.2
27
5760
XM_005261262.3
XP_005261319.1
27
5590
XM_011546137.2
XP_011544439.1
33
6109
XM_011546139.2
XP_011544441.1
33
6109
XM_011546142.2
XP_011544444.1
33
6109
XM_017028865.1
XP_016884354.1
33
6627
XM_017028866.1
XP_016884355.1
33
6433
XM_017028867.1
XP_016884356.1
33
7550
XM_017028868.1
XP_016884357.1
33
6279
XM_017028869.1
XP_016884358.1
33
6024
XM_017028870.1
XP_016884359.1
33
6025
XM_017028871.1
XP_016884360.1
33
6106
XM_017028872.1
XP_016884361.1
33
6109
XM_017028873.1
XP_016884362.1
32
6046
XM_017028874.1
XP_016884363.1
32
6025
XM_017028875.1
XP_016884364.1
32
6001
XM_017028876.1
XP_016884365.1
32
5971
XM_017028877.1
XP_016884366.1
31
5908
XM_017028878.1
XP_016884367.1
30
5794
XM_017028879.1
XP_016884368.1
28
5653
XM_017028880.1
XP_016884369.1
28
5976
XM_017028881.1
XP_016884370.1
28
5823
XM_017028882.1
XP_016884371.1
27
5590
XM_017028883.1
XP_016884372.1
32
5772
XM_017028884.1
XP_016884373.1
29
6102
XM_017028885.1
XP_016884374.1
13
3694
XM_017028886.1
XP_016884375.1
13
3542
GenBank mRNA
AB020626.1
-
7
2368
BC006562.2
-
2
534
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
exocytosis
cytoskeleton organization
actin filament depolymerization
oxidation-reduction process
actin binding
zinc ion binding
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, NAD(P)H as one donor, and incorporation of one atom of oxygen
Rab GTPase binding
FAD binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0