Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MICAL3 SIRNA Assays ›
          Gene Symbol:
          MICAL3
          Gene Name:
          microtubule associated monooxygenase, calponin and LIM domain containing 3
          Gene Aliases:
          MICAL-3
          Chromosome Location:
          Chr.22: 17787649 - 18024559 on Build GRCh38
          UniGene ID:
          Hs.528024
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s61183

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MICAL3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_015241.2 NP_056056.2 23, 24 3668
          XM_005261262.3 XP_005261319.1 23, 24 3498
          XM_011546137.2 XP_011544439.1 29, 30 4017
          XM_011546139.2 XP_011544441.1 29, 30 4017
          XM_011546142.2 XP_011544444.1 29, 30 4017
          XM_017028865.1 XP_016884354.1 29, 30 4535
          XM_017028866.1 XP_016884355.1 29, 30 4341
          XM_017028867.1 XP_016884356.1 29, 30 5458
          XM_017028868.1 XP_016884357.1 29, 30 4187
          XM_017028869.1 XP_016884358.1 29, 30 3932
          XM_017028870.1 XP_016884359.1 29, 30 3933
          XM_017028871.1 XP_016884360.1 29, 30 4014
          XM_017028872.1 XP_016884361.1 29, 30 4017
          XM_017028873.1 XP_016884362.1 28, 29 3954
          XM_017028874.1 XP_016884363.1 28, 29 3933
          XM_017028875.1 XP_016884364.1 28, 29 3909
          XM_017028876.1 XP_016884365.1 28, 29 3879
          XM_017028877.1 XP_016884366.1 27, 28 3816
          XM_017028879.1 XP_016884368.1 24, 25 3561
          XM_017028880.1 XP_016884369.1 24, 25 3884
          XM_017028881.1 XP_016884370.1 24, 25 3731
          XM_017028882.1 XP_016884371.1 23, 24 3498
          XM_017028883.1 XP_016884372.1 28, 29 3680
          XM_017028884.1 XP_016884373.1 25, 26 4010
          XM_017028885.1 XP_016884374.1 9, 10 1602
          XM_017028886.1 XP_016884375.1 9, 10 1450
          GenBank mRNA AB020626.1 - 3, 4 276
          CN425716.1 - 1, 2 63
          CR976532.1 - 2, 3 269

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          exocytosis
          cytoskeleton organization
          actin filament depolymerization
          oxidation-reduction process
          actin binding
          zinc ion binding
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, NAD(P)H as one donor, and incorporation of one atom of oxygen
          Rab GTPase binding
          FAD binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-9c8cf:80/100.66.75.150:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0