Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other OBSCN SIRNA Assays ›
          Gene Symbol:
          OBSCN
          Gene Name:
          obscurin, cytoskeletal calmodulin and titin-interacting RhoGEF
          Gene Aliases:
          ARHGEF30, UNC89
          Chromosome Location:
          Chr.1: 228208013 - 228381431 on Build GRCh38
          UniGene ID:
          Hs.650039
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s61234

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: OBSCN
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001098623.2 NP_001092093.2 86 19533
          NM_001271223.2 NP_001258152.2 97 22404
          XM_005273287.4 XP_005273344.1 98 22790
          XM_005273291.4 XP_005273348.1 98 22754
          XM_005273298.4 XP_005273355.1 97 22481
          XM_005273307.4 XP_005273364.1 93 21668
          XM_006711822.3 XP_006711885.1 96 22214
          XM_006711823.3 XP_006711886.1 96 22208
          XM_011544297.2 XP_011542599.1 90 20624
          XM_011544299.2 XP_011542601.1 89 20348
          XM_017002443.1 XP_016857932.1 98 22835
          XM_017002444.1 XP_016857933.1 98 22798
          XM_017002445.1 XP_016857934.1 98 22832
          XM_017002446.1 XP_016857935.1 98 22832
          XM_017002447.1 XP_016857936.1 98 22832
          XM_017002448.1 XP_016857937.1 98 22685
          XM_017002449.1 XP_016857938.1 97 22572
          XM_017002450.1 XP_016857939.1 97 22572
          XM_017002451.1 XP_016857940.1 97 22572
          XM_017002452.1 XP_016857941.1 97 22572
          XM_017002453.1 XP_016857942.1 97 22559
          XM_017002454.1 XP_016857943.1 97 22559
          XM_017002455.1 XP_016857944.1 97 22559
          XM_017002456.1 XP_016857945.1 97 22559
          XM_017002457.1 XP_016857946.1 97 22559
          XM_017002458.1 XP_016857947.1 96 22308
          XM_017002459.1 XP_016857948.1 96 22308
          XM_017002460.1 XP_016857949.1 96 22308
          XM_017002461.1 XP_016857950.1 96 22295
          XM_017002462.1 XP_016857951.1 96 22283
          XM_017002463.1 XP_016857952.1 96 22262
          XM_017002464.1 XP_016857953.1 96 22211
          XM_017002465.1 XP_016857954.1 95 22044
          XM_017002466.1 XP_016857955.1 95 22044
          XM_017002467.1 XP_016857956.1 95 22044
          XM_017002468.1 XP_016857957.1 94 21780
          XM_017002469.1 XP_016857958.1 92 21252
          XM_017002470.1 XP_016857959.1 87 19933
          GenBank mRNA AB046859.2 - 26 3308
          AM231061.1 - 12 1435
          DQ400343.1 - 12 1435

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          immunoglobulin

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          multicellular organism development
          regulation of Rho protein signal transduction
          protein localization to M-band
          positive regulation of apoptotic process
          positive regulation of GTPase activity
          sarcomere organization
          regulation of small GTPase mediated signal transduction
          mitophagy in response to mitochondrial depolarization
          protein serine/threonine kinase activity
          guanyl-nucleotide exchange factor activity
          Rho guanyl-nucleotide exchange factor activity
          protein binding
          calmodulin binding
          ATP binding
          structural constituent of muscle
          ankyrin binding
          titin binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline