siRNA Details
Gene Symbol: ROCK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005406.2 | NP_005397.1 | 31, 32 | 4801 | ||
XM_011526137.2 | XP_011524439.1 | 20, 21 | 3035 | |||
GenBank mRNA | BC113114.1 | - | 31, 32 | 3876 | 7961989 | |
U43195.1 | - | 31, 32 | 3860 |
Gene Symbol: ROCK1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033770.1 | 3, 4 | 737 | |||
GenBank mRNA | BC041849.1 | - | 3, 4 | 737 | ||
DB095042.1 | - | 3, 4 | 374 |

More Information
Other Species Targeted

Panther Classification:
