Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other KDM1A SIRNA Assays ›
          Gene Symbol:
          KDM1A
          Gene Name:
          lysine demethylase 1A
          Gene Aliases:
          AOF2, BHC110, CPRF, KDM1, LSD1
          Chromosome Location:
          Chr.1: 23019443 - 23083691 on Build GRCh38
          UniGene ID:
          Hs.591518
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s617
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 7,898.00
          Your Price
          Online offer:
          7898.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: KDM1A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001009999.2 NP_001009999.1 8, 9 1223
          NM_015013.3 NP_055828.2 7, 8 1163
          XM_005245786.2 XP_005245843.1 8, 9 1228
          XM_006710472.3 XP_006710535.1 7, 8 1168
          XM_006710473.2 XP_006710536.1 8, 9 1228
          XM_006710474.3 XP_006710537.1 8, 9 1228
          XM_017000716.1 XP_016856205.1 8, 9 1228
          XM_017000717.1 XP_016856206.1 7, 8 1168
          XM_017000718.1 XP_016856207.1 6, 7 634
          XM_017000719.1 XP_016856208.1 7, 8 778
          XM_017000720.1 XP_016856209.1 6, 7 748
          GenBank mRNA AB011173.1 - 7, 8 1116
          AK290328.1 - 7, 8 689
          AL833812.1 - 3, 4 285
          BC016639.1 - 5, 6 402
          BC040194.2 - 8, 9 1178 4815528
          BC048134.1 - 7, 8 1084 5298150

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00323448_m1

          Panther Classification:

          Molecular Function -

          oxidase

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          in utero embryonic development
          transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          protein demethylation
          blood coagulation
          cell proliferation
          regulation of double-strand break repair via homologous recombination
          regulation of primitive erythrocyte differentiation
          histone deacetylation
          pituitary gland development
          granulocyte differentiation
          negative regulation of protein binding
          histone H3-K9 demethylation
          positive regulation of histone ubiquitination
          cellular response to UV
          histone H3-K4 demethylation
          negative regulation of DNA binding
          negative regulation of sequence-specific DNA binding transcription factor activity
          negative regulation of DNA damage response, signal transduction by p53 class mediator
          positive regulation of erythrocyte differentiation
          positive regulation of megakaryocyte differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of hormone biosynthetic process
          positive regulation of sequence-specific DNA binding transcription factor activity
          negative regulation of histone H3-K4 methylation
          negative regulation of histone H3-K9 methylation
          muscle cell development
          oxidation-reduction process
          cellular response to gamma radiation
          negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          regulation of cellular protein localization
          positive regulation of neural precursor cell proliferation
          positive regulation of stem cell proliferation
          RNA polymerase II transcription factor binding
          p53 binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          histone deacetylase activity
          protein binding
          transcription factor binding
          oxidoreductase activity
          enzyme binding
          ligand-dependent nuclear receptor transcription coactivator activity
          demethylase activity
          histone demethylase activity
          histone demethylase activity (H3-K4 specific)
          histone demethylase activity (H3-K9 specific)
          histone demethylase activity (H3-dimethyl-K4 specific)
          telomeric DNA binding
          MRF binding
          transcription regulatory region DNA binding
          flavin adenine dinucleotide binding
          androgen receptor binding
          telomeric repeat-containing RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0