Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acadl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007381.4 | NP_031407.2 | 11 | 1309 | ||
GenBank mRNA | AK005140.1 | - | 11 | 1268 | ||
AK018319.1 | - | 10 | 1085 | |||
AK151845.1 | - | 11 | 1278 | |||
AK167537.1 | - | 11 | 1276 | |||
BC027412.1 | - | 11 | 1266 | 4236225 | ||
U21489.1 | - | 11 | 1249 |