Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NR3C1 SIRNA Assays ›
Gene Symbol:
NR3C1
Gene Name:
nuclear receptor subfamily 3 group C member 1
Gene Aliases:
GCCR, GCR, GCRST, GR, GRL
Chromosome Location:
Chr.5: 143277931 - 143435512 on Build GRCh38
UniGene ID:
Hs.122926
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s6186
Validated
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NR3C1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000176.2
NP_000167.1
2
1532
NM_001018074.1
NP_001018084.1
2
1362
NM_001018075.1
NP_001018085.1
2
1265
NM_001018076.1
NP_001018086.1
2
1158
NM_001018077.1
NP_001018087.1
2
2034
NM_001020825.1
NP_001018661.1
2
1532
NM_001024094.1
NP_001019265.1
2
1532
NM_001204258.1
NP_001191187.1
2
1532
NM_001204259.1
NP_001191188.1
2
1532
NM_001204260.1
NP_001191189.1
2
1532
NM_001204261.1
NP_001191190.1
2
1532
NM_001204262.1
NP_001191191.1
2
1532
NM_001204263.1
NP_001191192.1
2
1532
NM_001204264.1
NP_001191193.1
2
1532
NM_001204265.1
NP_001191194.1
2
1532
XM_005268419.3
XP_005268476.1
2
1262
XM_005268420.4
XP_005268477.1
2
1443
XM_005268422.3
XP_005268479.1
2
1324
XM_005268423.3
XP_005268480.1
2
1313
XM_017009397.1
XP_016864886.1
2
1158
XM_017009398.1
XP_016864887.1
2
1457
GenBank mRNA
AB307716.1
-
1
1040
AK223594.1
-
2
1386
AK314789.1
-
2
1161
AM183262.1
-
2
975
BC015610.2
-
2
1480
4810424
BX640610.1
-
2
1223
BX648553.1
-
3
1324
HQ450643.1
-
1
1040
HQ450644.1
-
1
1040
HQ692822.1
-
1
1040
JN797823.1
-
1
1039
JN797824.1
-
1
1040
KU921432.1
-
1
1053
U01351.1
-
2
1533
X03225.1
-
2
1172
X03348.1
-
2
1172
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00230813_m1
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
apoptotic process
chromosome segregation
mitotic nuclear division
signal transduction
covalent chromatin modification
glucocorticoid receptor signaling pathway
glucocorticoid mediated signaling pathway
positive regulation of transcription from RNA polymerase II promoter
cell division
cellular response to steroid hormone stimulus
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
RNA binding
glucocorticoid receptor activity
steroid binding
protein binding
zinc ion binding
protein complex binding
glucocorticoid-activated RNA polymerase II transcription factor binding transcription factor activity
steroid hormone binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-9fjjm:80/10.239.165.64:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0