Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ADAM17 SIRNA Assays ›
          Gene Symbol:
          Adam17
          Gene Name:
          a disintegrin and metallopeptidase domain 17
          Gene Aliases:
          CD156b, Tace
          Chromosome Location:
          Chr.12: 21323509 - 21373632 on Build GRCm38
          UniGene ID:
          Mm.27681
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s61957

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Adam17
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001277266.1 NP_001264195.1 5 669
          NM_001291871.1 NP_001278800.1 5 669
          NM_009615.6 NP_033745.4 5 669
          GenBank mRNA AB021709.1 - 5 630
          AJ007365.1 - 5 469
          AK137693.1 - 5 576
          AK139471.1 - 5 575
          AK142441.1 - 5 609
          AK144161.1 - 5 599
          AK149288.1 - 5 572
          AK149614.1 - 5 576
          BC026052.1 - 4 432
          BC048792.1 - 5 577
          BC094655.1 - 5 583 5705503
          BC114999.1 - 5 529 6506501
          BC138416.1 - 5 548 8861436
          BC138420.1 - 5 548 8861440
          BC145270.1 - 5 548 9053824
          CA325381.1 - 5 637
          U69613.1 - 5 537
          U69614.1 - 5 621

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          positive regulation of protein phosphorylation
          germinal center formation
          proteolysis
          membrane protein ectodomain proteolysis
          cell adhesion
          epidermal growth factor receptor signaling pathway
          Notch signaling pathway
          Notch receptor processing
          positive regulation of cell proliferation
          positive regulation of T cell chemotaxis
          B cell differentiation
          positive regulation of cell growth
          positive regulation of cell migration
          positive regulation of transforming growth factor beta receptor signaling pathway
          negative regulation of transforming growth factor beta receptor signaling pathway
          positive regulation of cyclin-dependent protein serine/threonine kinase activity involved in G1/S transition of mitotic cell cycle
          response to lipopolysaccharide
          positive regulation of chemokine production
          regulation of mast cell apoptotic process
          T cell differentiation in thymus
          cell adhesion mediated by integrin
          epidermal growth factor-activated receptor transactivation by G-protein coupled receptor signaling pathway
          response to drug
          negative regulation of apoptotic process
          positive regulation of epidermal growth factor-activated receptor activity
          spleen development
          cell motility
          defense response to Gram-positive bacterium
          PMA-inducible membrane protein ectodomain proteolysis
          positive regulation of cellular component movement
          response to high density lipoprotein particle
          metalloendopeptidase activity
          Notch binding
          interleukin-6 receptor binding
          integrin binding
          protein binding
          peptidase activity
          metallopeptidase activity
          zinc ion binding
          hydrolase activity
          SH3 domain binding
          PDZ domain binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0