Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ADRA2A SIRNA Assays ›
          Gene Symbol:
          Adra2a
          Gene Name:
          adrenergic receptor, alpha 2a
          Gene Aliases:
          AW122659, Adra-2, Adra-2a, alpha(2A)AR, alpha2-C10, alpha2A, alpha2A-AR
          Chromosome Location:
          Chr.19: 54045182 - 54048982 on Build GRCm38
          UniGene ID:
          Mm.235195
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s62068

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Adra2a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_007417.4 NP_031443.4 1 1235
          GenBank mRNA AK046802.1 - 2 1029
          AK140461.1 - 2 1029
          AK141573.1 - 1 1188
          BC138531.1 - 1 851 8861551

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_012739, U79031.

          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of cytokine production
          acute inflammatory response
          DNA replication
          G-protein coupled receptor signaling pathway
          negative regulation of adenylate cyclase activity
          cell-cell signaling
          female pregnancy
          negative regulation of norepinephrine secretion
          positive regulation of cell migration
          negative regulation of cAMP biosynthetic process
          activation of protein kinase activity
          activation of protein kinase B activity
          cellular response to hormone stimulus
          epidermal growth factor-activated receptor transactivation by G-protein coupled receptor signaling pathway
          glucose homeostasis
          fear response
          positive regulation of MAP kinase activity
          positive regulation of epidermal growth factor-activated receptor activity
          positive regulation of vasodilation
          negative regulation of calcium ion-dependent exocytosis
          negative regulation of insulin secretion
          thermoception
          negative regulation of lipid catabolic process
          positive regulation of membrane protein ectodomain proteolysis
          negative regulation of insulin secretion involved in cellular response to glucose stimulus
          negative regulation of uterine smooth muscle contraction
          adrenergic receptor signaling pathway
          adenylate cyclase-inhibiting adrenergic receptor signaling pathway
          phospholipase C-activating adrenergic receptor signaling pathway
          activation of MAPK activity by adrenergic receptor signaling pathway
          positive regulation of wound healing
          adrenergic receptor activity
          alpha2-adrenergic receptor activity
          protein kinase binding
          alpha-2C adrenergic receptor binding
          thioesterase binding
          heterotrimeric G-protein binding
          protein homodimerization activity
          protein heterodimerization activity
          epinephrine binding
          norepinephrine binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0