Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr2f2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009697.3 | NP_033827.2 | 2 | 1123 | ||
NM_183261.3 | NP_899084.2 | 2 | 1071 | |||
GenBank mRNA | AK034619.1 | - | 1 | 242 | ||
AK134384.1 | - | 1 | 532 | |||
AK140125.1 | - | 2 | 1047 | |||
BC042484.1 | - | 2 | 1123 | 5368085 | ||
BC094360.1 | - | 2 | 1048 | |||
U07635.1 | - | 2 | 1006 | |||
X76653.1 | - | 2 | 894 |