Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007483.2 | NP_031509.1 | 1 | 465 | ||
GenBank mRNA | AF481943.1 | - | 1 | 112 | ||
AK086873.1 | - | 1 | 490 | |||
AK164518.1 | - | 1 | 490 | |||
AK167917.1 | - | 1 | 490 | |||
BC018275.1 | - | 1 | 465 | 4168565 | ||
BC023334.1 | - | 1 | 221 | |||
BC110992.1 | - | 1 | 476 | 5376565 | ||
CT010379.1 | - | 1 | 112 |