siRNA Details
Gene Symbol: Atp1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009721.6 | NP_033851.1 | 1, 2 | 587 | ||
GenBank mRNA | AK005071.1 | - | 1, 2 | 234 | ||
AK010677.1 | - | 1, 2 | 588 | |||
AK140862.1 | - | 1, 2 | 234 | |||
AK160405.1 | - | 1, 2 | 233 | |||
AK161891.1 | - | 1, 2 | 238 | |||
BC027319.1 | - | 1, 2 | 226 | 2648940 | ||
BC094070.1 | - | 1, 2 | 225 | 4974089 | ||
X16646.1 | - | 1, 2 | 124 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
