Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BAK1 SIRNA Assays ›
          Gene Symbol:
          Bak1
          Gene Name:
          BCL2-antagonist/killer 1
          Gene Aliases:
          Bak, N-BAK1, N-Bak
          Chromosome Location:
          Chr.17: 27019812 - 27028839 on Build GRCm38
          UniGene ID:
          Mm.2443
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s62861

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Bak1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_007523.2 NP_031549.2 5 734
          GenBank mRNA AF402617.1 - 5 501
          AK089220.1 - 5 672
          BC057589.1 - 6 706 6411580
          CX564692.1 - 5 667
          Y13231.1 - 5 732

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          leukocyte homeostasis
          B cell homeostasis
          B cell apoptotic process
          release of cytochrome c from mitochondria
          blood vessel remodeling
          myeloid cell homeostasis
          B cell negative selection
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          mitochondrial fusion
          cell proliferation
          negative regulation of cell proliferation
          intrinsic apoptotic signaling pathway in response to DNA damage
          activation of cysteine-type endopeptidase activity involved in apoptotic process by cytochrome c
          response to fungus
          response to mycotoxin
          establishment or maintenance of transmembrane electrochemical gradient
          response to gamma radiation
          positive regulation of calcium ion transport into cytosol
          negative regulation of gene expression
          cytolysis
          negative regulation of endoplasmic reticulum calcium ion concentration
          negative regulation of peptidyl-serine phosphorylation
          cellular response to UV
          limb morphogenesis
          regulation of apoptotic process
          positive regulation of apoptotic process
          regulation of protein homodimerization activity
          regulation of protein heterodimerization activity
          fibroblast apoptotic process
          positive regulation of proteolysis
          regulation of mitochondrial membrane permeability
          post-embryonic camera-type eye morphogenesis
          homeostasis of number of cells
          regulation of cell cycle
          regulation of mitochondrial membrane potential
          vagina development
          intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
          thymocyte apoptotic process
          positive regulation of release of cytochrome c from mitochondria
          apoptotic signaling pathway
          extrinsic apoptotic signaling pathway in absence of ligand
          activation of cysteine-type endopeptidase activity
          positive regulation of endoplasmic reticulum unfolded protein response
          apoptotic process involved in patterning of blood vessels
          protein binding
          heat shock protein binding
          identical protein binding
          protein homodimerization activity
          metal ion binding
          protein heterodimerization activity
          chaperone binding
          BH domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0