siRNA Details
Gene Symbol: Bcan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007529.2 | NP_031555.2 | 12 | 2609 | ||
GenBank mRNA | AK144405.1 | - | 12 | 2410 | ||
AK163646.1 | - | 5 | 670 | |||
BC052032.1 | - | 12 | 2404 | 5695027 | ||
X87096.1 | - | 12 | 2485 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
