Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BCL2L1 SIRNA Assays ›
          Gene Symbol:
          Bcl2l1
          Gene Name:
          BCL2-like 1
          Gene Aliases:
          Bcl(X)L, Bcl-XL, Bcl2l, BclX, bcl-x, bcl2-L-1
          Chromosome Location:
          Chr.2: 152758732 - 152832581 on Build GRCm38
          UniGene ID:
          Mm.238213
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s62925

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Bcl2l1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001289716.1 NP_001276645.1 2 527
          NM_001289717.1 NP_001276646.1 2 372
          NM_001289739.1 NP_001276668.1 2 527
          NM_009743.5 NP_033873.3 2 321
          GenBank mRNA AK146461.1 - 2 477
          AK150028.1 - 2 263
          AK172250.1 - 2 288
          BB856100.1 - 2 324
          BC089016.1 - 2 275 30606568
          BC089017.1 - 1 112 30605282
          L35049.1 - 2 325
          U10100.1 - 1 83
          U10101.1 - 1 83
          U10102.1 - 2 320
          U51278.1 - 1 185
          U51279.1 - 2 318
          X83574.1 - 1 83

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          cytokinesis
          ovarian follicle development
          in utero embryonic development
          release of cytochrome c from mitochondria
          response to ischemia
          apoptotic process
          mitotic cell cycle checkpoint
          germ cell development
          spermatogenesis
          cell proliferation
          positive regulation of cell proliferation
          male gonad development
          intrinsic apoptotic signaling pathway in response to DNA damage
          response to radiation
          fertilization
          response to virus
          suppression by virus of host apoptotic process
          response to cytokine
          growth
          regulation of apoptotic process
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          negative regulation of neuron apoptotic process
          response to cycloheximide
          regulation of mitochondrial membrane permeability
          neuron apoptotic process
          regulation of mitochondrial membrane potential
          cellular process regulating host cell cycle in response to virus
          mitochondrion morphogenesis
          cellular response to amino acid stimulus
          cellular response to alkaloid
          cellular response to gamma radiation
          apoptotic process in bone marrow
          negative regulation of establishment of protein localization to plasma membrane
          negative regulation of release of cytochrome c from mitochondria
          extrinsic apoptotic signaling pathway in absence of ligand
          hepatocyte apoptotic process
          negative regulation of execution phase of apoptosis
          negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
          negative regulation of anoikis
          negative regulation of intrinsic apoptotic signaling pathway
          protein binding
          protein kinase binding
          identical protein binding
          protein homodimerization activity
          cysteine-type endopeptidase inhibitor activity involved in apoptotic process
          protein heterodimerization activity
          BH domain binding
          BH3 domain binding
          MDM2/MDM4 family protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-mc8hj:80/100.66.75.242:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0