siRNA Details
Gene Symbol: Caml | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007596.2 | NP_031622.2 | 2 | 539 | ||
GenBank mRNA | AK002895.1 | - | 2 | 537 | ||
AK218385.1 | - | 1 | 32 | |||
BC005688.1 | - | 2 | 462 | 3156307 | ||
CF915604.1 | - | 2 | 560 | |||
U21960.1 | - | 2 | 487 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
