Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007651.3 | NP_031677.1 | 5 | 570 | ||
GenBank mRNA | AA177409.1 | - | 5 | 458 | ||
AK156510.1 | - | 5 | 477 | |||
AK160326.1 | - | 5 | 468 | |||
BC021310.1 | - | 5 | 484 | 5010267 | ||
BC052905.1 | - | 5 | 435 | 30078163 | ||
X97227.1 | - | 4 | 371 | |||
Z16071.1 | - | 4 | 356 |