Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GYS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161587.1 | NP_001155059.1 | 9 | 1308 | ||
NM_002103.4 | NP_002094.2 | 10 | 1500 | |||
NR_027763.1 | 9 | 1318 | ||||
GenBank mRNA | AK223579.1 | - | 10 | 1456 | ||
AK291672.1 | - | 10 | 1456 | |||
AK294349.1 | - | 5 | 416 | |||
AK300369.1 | - | 10 | 1288 | |||
AK302738.1 | - | 9 | 1274 | |||
AK303712.1 | - | 8 | 1156 | |||
BC002617.2 | - | 10 | 1439 | 3143019 | ||
BC003182.1 | - | 9 | 1265 | 2958297 | ||
BC007688.2 | - | 10 | 1426 | 3953692 | ||
J04501.1 | - | 10 | 1419 | |||
U32573.1 | - | 10 | 1401 |