Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009900.2 | NP_034030.2 | 8 | 1476 | ||
GenBank mRNA | AF097415.1 | - | 8 | 954 | ||
AF139724.1 | - | 8 | 905 | |||
AK141656.1 | - | 9 | 2112 | |||
AK155104.1 | - | 8 | 770 | |||
AK165000.1 | - | 8 | 1004 | |||
AK171231.1 | - | 8 | 770 | |||
AK172148.1 | - | 5 | 1459 | |||
BC120699.1 | - | 8 | 867 | 40129622 | ||
BC137625.1 | - | 8 | 867 | 8860644 |