Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cobl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282993.1 | NP_001269922.1 | 11 | 2427 | ||
NM_001282994.1 | NP_001269923.1 | 12 | 2333 | |||
NM_172496.3 | NP_766084.3 | 12 | 2502 | |||
GenBank mRNA | AK122334.1 | - | 12 | 2333 | ||
AK147366.1 | - | 11 | 2427 | |||
AK147633.1 | - | 11 | 2427 | |||
AY308745.1 | - | 12 | 2342 | |||
BC060061.1 | - | 1 | 66 | |||
BC145116.1 | - | 9 | 1807 | |||
BC145118.1 | - | 12 | 2342 | 9053666 | ||
BC150706.1 | - | 12 | 2342 | 9087617 |