Search
Search

HAS1| Gene Symbol: HAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297436.1 | NP_001284365.1 | 4, 5 | 1110 | ||
| NM_001523.3 | NP_001514.2 | 4, 5 | 1113 | |||
| GenBank mRNA | AY916556.1 | - | 3 | 410 | ||
| BC035837.1 | - | 4, 5 | 1092 | 5589083 | ||
| D84424.1 | - | 3, 4 | 1097 | |||
| U59269.1 | - | 4, 5 | 1089 | |||