Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007802.4 | NP_031828.2 | 4 | 490 | ||
GenBank mRNA | AK003425.1 | - | 4 | 415 | ||
AK132648.1 | - | 4 | 420 | |||
BC046320.1 | - | 4 | 444 | 6390854 | ||
X94444.1 | - | 3 | 362 |