siRNA Details
Gene Symbol: Cycs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007808.4 | NP_031834.1 | 3 | 314 | ||
GenBank mRNA | AK002732.1 | - | 3 | 316 | ||
AK003168.1 | - | 3 | 343 | |||
AK005336.1 | - | 3 | 386 | |||
AK010651.1 | - | 3 | 318 | |||
AK088098.1 | - | 3 | 315 | |||
AK143887.1 | - | 3 | 315 | |||
AK168017.1 | - | 3 | 315 | |||
BC034363.1 | - | 3 | 298 | 2650742 | ||
BC092213.1 | - | 2 | 263 | 6845432 | ||
BC094054.1 | - | 3 | 303 | 2865963 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
