Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dclk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111051.1 | NP_001104521.1 | 3 | 478 | ||
NM_001111052.1 | NP_001104522.1 | 3 | 478 | |||
NM_001195538.1 | NP_001182467.1 | 7 | 1339 | |||
NM_001195539.1 | NP_001182468.1 | 3 | 478 | |||
NM_019978.3 | NP_064362.1 | 7 | 1339 | |||
GenBank mRNA | AB093234.1 | - | 7 | 1296 | ||
AF155819.1 | - | 7 | 1237 | |||
AF155820.1 | - | 7 | 1278 | |||
AF155821.1 | - | 3 | 337 | |||
AK032424.1 | - | 3 | 320 | |||
AK081200.1 | - | 3 | 321 | |||
AK163437.1 | - | 1 | 696 | |||
AY968047.1 | - | 3 | 229 | |||
AY968048.1 | - | 3 | 229 | |||
BC133685.1 | - | 7 | 1141 | 40110683 |