Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019910.2 | NP_064294.2 | 4 | 364 | ||
GenBank mRNA | BC005655.1 | - | 361 | 3709995 | ||
BC024627.1 | - | 338 | 4925312 | |||
BC045522.1 | - | 4 | 348 | 4190591 | ||
BC090399.1 | - | 338 | 4168483 | |||
BY150555.1 | - | 4 | 363 | |||
DQ237936.1 | - | 356 | ||||
S76879.1 | - | 397 | ||||
U03711.1 | - | 361 |
Gene Symbol: Dcpp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC059096.1 | - | 3 | 378 | 4914089 | |
BC171996.1 | - | 360 | 9054282 | |||
DQ237937.1 | - | 359 |