Search
Search

Dlg4| Gene Symbol: Dlg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109752.1 | NP_001103222.1 | 7 | 877 | ||
| NM_007864.3 | NP_031890.1 | 7 | 886 | |||
| GenBank mRNA | BC014807.1 | - | 7 | 622 | 4501403 | |
| BI738861.1 | - | 7 | 634 | |||
| BQ959940.1 | - | 1 | 29 | |||
| D50621.1 | - | 7 | 628 | |||