Search
Search

Dync1i2| Gene Symbol: Dync1i2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198872.1 | NP_001185801.1 | 11 | 1042 | ||
| NM_001198873.1 | NP_001185802.1 | 11 | 1027 | |||
| NM_001198874.1 | NP_001185803.1 | 9 | 973 | |||
| NM_001198875.1 | NP_001185804.1 | 9 | 931 | |||
| NM_001198876.1 | NP_001185805.1 | 8 | 913 | |||
| NM_001198877.1 | NP_001185806.1 | 11 | 1031 | |||
| NM_001198878.1 | NP_001185807.1 | 11 | 1016 | |||
| NM_010064.4 | NP_034194.1 | 8 | 902 | |||
| GenBank mRNA | AF063231.1 | - | 8 | 841 | ||
| AK088015.1 | - | 8 | 873 | |||
| AK141837.1 | - | 9 | 941 | |||
| AK150173.1 | - | 8 | 863 | |||
| AK161831.1 | - | 4 | 1563 | |||
| AK164322.1 | - | 9 | 897 | |||
| AK164437.1 | - | 8 | 837 | |||
| AK168730.1 | - | 8 | 877 | |||
| BC139374.1 | - | 8 | 797 | 8862396 | ||
| BC139375.1 | - | 8 | 797 | 8862397 | ||
| GU992208.1 | - | 11 | 933 | |||
| GU992209.1 | - | 11 | 918 | |||
| GU992210.1 | - | 9 | 822 | |||
| GU992211.1 | - | 11 | 971 | |||
| GU992212.1 | - | 11 | 956 | |||