Search
Search

Dst| Gene Symbol: Dst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276764.1 | NP_001263693.1 | 20 | 2715 | ||
| NM_010081.2 | NP_034211.2 | 7 | 1156 | |||
| NM_133833.3 | NP_598594.2 | 17 | 2134 | |||
| NM_134448.3 | NP_604443.2 | 17 | 2134 | |||
| GenBank mRNA | AF396877.1 | - | 7 | 1147 | ||
| AF396878.1 | - | 17 | 2146 | |||
| AF396879.1 | - | 17 | 2146 | |||
| AK141319.1 | - | 1 | 2222 | |||