Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Edil3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037987.3 | NP_001033076.1 | 5 | 922 | ||
NM_010103.4 | NP_034233.1 | 4 | 892 | |||
GenBank mRNA | AF031524.1 | - | 5 | 1030 | ||
AF031525.1 | - | 5 | 961 | |||
AK036117.1 | - | 5 | 883 | |||
AK046885.1 | - | 5 | 923 | |||
AK081003.1 | - | 4 | 724 | |||
AK084215.1 | - | 5 | 777 | |||
AK135220.1 | - | 5 | 886 | |||
AK139093.1 | - | 5 | 884 | |||
AK141907.1 | - | 5 | 906 | |||
AK157810.1 | - | 5 | 907 | |||
BC056386.1 | - | 5 | 822 | 6854497 |