Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk1b9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010116.1 | NP_034246.1 | 278 | |||
GenBank mRNA | BC024624.1 | - | 257 | 4914236 | ||
BC048869.1 | - | 242 | 4164261 | |||
M17962.1 | - | 278 |