Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HLA-B SIRNA Assays ›
          Gene Symbol:
          HLA-B
          Gene Name:
          major histocompatibility complex, class I, B
          Gene Aliases:
          AS, B-4901, B-5001, Bw-47, Bw-50, HEL-S-83, HLA-B*45ZJ, HLA-B-3506, HLA-B-3905, HLA-B-5502, HLA-B-5602, HLA-B15, HLA-B39, HLA-B49, HLA-B50, HLA-B55, HLA-B59, HLA-B61, HLA-Cw, HLA-DRB1, HLAB, SPDA1
          Chromosome Location:
          Chr.6: 31353866 - 31357245 on Build GRCh38
          UniGene ID:
          Hs.654404
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s6572

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HLA-B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005514.7 NP_005505.2 4 876
          XM_011514557.1 XP_011512859.1 4 799
          GenBank mRNA AB008102.1 - 4 789
          AF006634.1 - 4 789
          AF016641.1 - 4 789
          AF022783.1 - 4 789
          AF026218.1 - 4 789
          AF130734.1 - 5 789
          AF170577.1 - 4 789
          AF189017.1 - 4 789
          AF389378.1 - 4 793
          AJ131118.1 - 4 789
          AJ251593.1 - 4 788
          AJ506054.1 - 1 170
          AJ535113.1 - 4 793
          AJ634528.1 - 4 789
          AK026595.1 - 5 812
          AK124160.1 - 4 1059
          AK129796.1 - 4 810
          AK130137.1 - 4 810
          AK130155.1 - 4 812
          AK130166.1 - 4 810
          AK130169.1 - 4 816
          AK130654.1 - 5 749
          AK290570.1 - 4 810
          AK292226.1 - 4 810
          AK300685.1 - 5 719
          AK313911.1 - 4 810
          AM711637.1 - 4 789
          AY007140.1 - 4 809
          AY221104.1 - 3 693
          AY569160.1 - 3 716
          BC007243.1 - 5 783
          BC013187.1 - 4 798 4344323
          BC091497.1 - 4 793 30386550
          BX647315.1 - 5 811
          D14343.1 - 4 753
          D25275.1 - 4 789
          D31816.1 - 4 789
          D49824.1 - 4 789
          D50290.1 - 4 789
          D50291.1 - 4 789
          D50292.1 - 4 789
          D50293.1 - 4 789
          D50294.1 - 4 789
          D50295.1 - 4 789
          D50296.1 - 4 789
          D50299.1 - 4 789
          D50300.1 - 4 789
          D83043.1 - 4 789
          D85761.1 - 5 789
          D85762.1 - 4 789
          D87665.1 - 5 780
          DQ336696.1 - 4 777
          DQ336697.1 - 4 783
          DQ336698.1 - 4 777
          EU048339.1 - 1 170
          EU668336.1 - 4 789
          FM205711.1 - 4 789
          HE663398.1 - 5 738
          HG327115.1 - 4 789
          HG327117.1 - 4 789
          HG327119.1 - 4 789
          HG327120.1 - 4 789
          HG514457.1 - 4 789
          KU204955.1 - 2 446
          L04695.1 - 4 789
          L04696.1 - 4 789
          L07743.1 - 4 789
          L07950.1 - 4 789
          L09735.1 - 4 789
          L09736.1 - 4 789
          L11570.1 - 4 789
          L11571.1 - 4 789
          L11603.1 - 4 789
          L11604.1 - 4 789
          L11666.1 - 4 789
          L15005.1 - 4 789
          L19923.1 - 4 789
          L19937.1 - 4 789
          L20089.1 - 4 784
          L20090.1 - 5 784
          L22027.1 - 4 789
          L32862.1 - 4 789
          L33922.1 - 4 789
          L37880.1 - 4 789
          L37881.1 - 4 789
          L38504.1 - 4 789
          L41214.1 - 5 789
          L41925.1 - 5 789
          L42345.1 - 4 789
          M12678.1 - 4 780
          M15470.1 - 4 780
          M16102.1 - 4 796
          M24033.1 - 4 789
          M27540.1 - 3 603
          M28203.1 - 4 778
          M28204.1 - 4 753
          M28205.1 - 5 789
          M32317.1 - 4 789
          M32318.1 - 5 789
          M32319.1 - 5 789
          M32320.1 - 4 789
          M62852.1 - 3 717
          M75138.1 - 4 789
          M77774.1 - 5 789
          M77775.1 - 4 789
          M77776.1 - 4 789
          M77777.1 - 4 789
          M77778.1 - 4 789
          M81798.1 - 4 789
          M83191.1 - 3 717
          M83192.1 - 3 717
          M83193.1 - 3 717
          M83194.1 - 3 717
          M83195.1 - 3 717
          M84380.1 - 4 789
          M84381.1 - 4 789
          M84382.1 - 4 789
          M84383.1 - 4 789
          M84384.1 - 4 789
          M84385.1 - 4 789
          M84694.1 - 4 789
          M86403.1 - 4 765
          U01848.1 - 4 789
          U03027.1 - 4 789
          U03698.1 - 4 789
          U03859.1 - 4 789
          U04244.1 - 4 789
          U04245.1 - 4 789
          U06697.1 - 3 716
          U06862.1 - 4 789
          U09864.1 - 4 789
          U09912.1 - 4 789
          U11261.1 - 4 789
          U11262.1 - 4 789
          U11263.1 - 4 789
          U11264.1 - 4 789
          U11265.1 - 4 789
          U11266.1 - 4 789
          U11267.1 - 4 789
          U11268.1 - 4 789
          U11269.1 - 4 789
          U14943.1 - 4 789
          U17107.1 - 4 789
          U17572.1 - 4 789
          U21052.1 - 4 789
          U21053.1 - 4 789
          U29057.1 - 4 789
          U30904.1 - 4 789
          U30936.1 - 4 789
          U31971.1 - 4 789
          U35431.1 - 4 789
          U35734.1 - 4 789
          U36392.1 - 4 789
          U36492.1 - 4 789
          U38800.1 - 4 789
          U50710.1 - 4 789
          U57966.1 - 4 789
          U63653.1 - 4 789
          U64801.1 - 4 789
          U70528.1 - 4 789
          U70529.1 - 4 789
          U75533.1 - 4 789
          U80945.1 - 4 789
          U88407.1 - 4 789
          V00527.1 - 1 52
          X61706.1 - 4 789
          X61708.1 - 5 789
          X61709.1 - 4 789
          X61710.1 - 4 789
          X64366.1 - 4 789
          X64454.1 - 4 789
          X73578.1 - 4 789
          X75953.1 - 5 789
          X81363.1 - 4 789
          X84725.1 - 4 789
          X90390.1 - 4 789
          X90391.1 - 4 789
          X91749.1 - 4 789
          Y08994.1 - 5 789
          Y08995.1 - 4 789
          Y09118.1 - 4 789
          Y13567.1 - 4 789
          Z15143.1 - 5 789
          Z22651.1 - 4 789
          Z33453.1 - 4 790

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          Gene Ontology Categories:

          Function(s) Process(es)

          antigen processing and presentation of peptide antigen via MHC class I
          antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent
          antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-independent
          viral process
          antigen processing and presentation
          regulation of immune response
          interferon-gamma-mediated signaling pathway
          type I interferon signaling pathway
          antigen binding
          peptide antigen binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0