Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 企業服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other F2RL1 SIRNA Assays ›
          Gene Symbol:
          F2rl1
          Gene Name:
          coagulation factor II (thrombin) receptor-like 1
          Gene Aliases:
          Gpcr11, PAR-2, Par2
          Chromosome Location:
          Chr.13: 95511718 - 95525240 on Build GRCm38
          UniGene ID:
          Mm.1614
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s65793

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: F2rl1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_007974.4 NP_032000.3 2 1054
          GenBank mRNA AK144986.1 - 2 1038
          AK158462.1 - 2 1042
          AK160916.1 - 2 1042
          AX785009.1 - 2 1013
          BC025432.1 - 2 1039 5006769
          Z48043.1 - 2 1013

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          T cell activation involved in immune response
          immune system process
          positive regulation of leukocyte chemotaxis
          positive regulation of cytokine secretion involved in immune response
          positive regulation of glomerular filtration
          inflammatory response
          signal transduction
          G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          blood coagulation
          negative regulation of tumor necrosis factor-mediated signaling pathway
          positive regulation of phosphatidylinositol 3-kinase signaling
          regulation of blood coagulation
          positive regulation of cell migration
          positive regulation of actin filament depolymerization
          positive regulation of pseudopodium assembly
          positive regulation of superoxide anion generation
          positive regulation of toll-like receptor 2 signaling pathway
          negative regulation of toll-like receptor 3 signaling pathway
          positive regulation of toll-like receptor 3 signaling pathway
          positive regulation of toll-like receptor 4 signaling pathway
          positive regulation of Rho protein signal transduction
          chemokine (C-C motif) ligand 2 secretion
          neutrophil activation
          regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of eosinophil degranulation
          innate immune response
          positive regulation of vasodilation
          positive regulation of transcription from RNA polymerase II promoter
          regulation of JNK cascade
          negative regulation of JNK cascade
          positive regulation of JNK cascade
          interleukin-1 beta secretion
          leukocyte migration
          positive regulation of chemotaxis
          positive regulation of positive chemotaxis
          defense response to virus
          positive regulation of phagocytosis, engulfment
          establishment of endothelial barrier
          positive regulation of ERK1 and ERK2 cascade
          thrombin receptor signaling pathway
          leukocyte proliferation
          positive regulation of neutrophil mediated killing of gram-negative bacterium
          interleukin-10 secretion
          interferon-gamma secretion
          chemokine secretion
          negative regulation of chemokine secretion
          mature conventional dendritic cell differentiation
          positive regulation of renin secretion into blood stream
          positive regulation of interleukin-8 secretion
          positive regulation of interleukin-6 secretion
          G-protein alpha-subunit binding
          signal transducer activity
          G-protein coupled receptor activity
          thrombin receptor activity
          G-protein beta-subunit binding
          heterotrimeric G-protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-gh6bl:80/100.66.76.225:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0