Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3pxd2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164717.1 | NP_001158189.1 | 10 | 1075 | ||
NM_008018.4 | NP_032044.2 | 11 | 1159 | |||
GenBank mRNA | AJ007012.1 | - | 11 | 1159 | ||
AK085227.1 | - | 4 | 998 | |||
BC118022.1 | - | 10 | 911 | 40097448 |