Search
Search

Folr1| Gene Symbol: Folr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252552.1 | NP_001239481.1 | 6 | 935 | ||
| NM_001252553.1 | NP_001239482.1 | 6 | 861 | |||
| NM_001252554.1 | NP_001239483.1 | 6 | 848 | |||
| NM_008034.3 | NP_032060.2 | 6 | 932 | |||
| GenBank mRNA | AF096319.1 | - | 6 | 827 | ||
| AK002275.1 | - | 6 | 927 | |||
| BC002071.1 | - | 3 | 501 | |||
| BC037078.1 | - | 5 | 673 | |||
| BC138781.1 | - | 5 | 565 | 8861803 | ||
| BC138795.1 | - | 5 | 652 | 8861817 | ||
| BC145414.1 | - | 5 | 652 | 9053972 | ||
| M64782.1 | - | 3 | 501 | |||