Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008069.4 | NP_032095.1 | 7 | 731 | ||
GenBank mRNA | BC130258.1 | - | 7 | 735 | 40142758 | |
BC157920.1 | - | 7 | 797 | 9007350 |