siRNA Details
Gene Symbol: Gbx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010262.3 | NP_034392.1 | 2 | 983 | ||
GenBank mRNA | AB221564.1 | - | 2 | 562 | ||
AK138361.1 | - | 1 | 84 | |||
AK141657.1 | - | 2 | 299 | |||
BC050989.1 | - | 3 | 897 | |||
BC116965.1 | - | 2 | 609 | 40126284 | ||
BC120492.1 | - | 2 | 609 | 40129415 | ||
L39770.1 | - | 2 | 957 | |||
Z48800.1 | - | 2 | 983 |

More Information
Other Species Targeted

Panther Classification:
