Search
Search

Gng5| Gene Symbol: Gng5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010318.2 | NP_034448.2 | 3 | 441 | ||
| GenBank mRNA | AK146065.1 | - | 3 | 364 | ||
| BC002316.1 | - | 3 | 392 | 3593324 | ||
| BC010725.1 | - | 2 | 2321 | |||
| BC087872.1 | - | 3 | 363 | 6582824 | ||