Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GPX1 SIRNA Assays ›
          Gene Symbol:
          Gpx1
          Gene Name:
          glutathione peroxidase 1
          Gene Aliases:
          AI195024, AL033363, CGPx, GPx-1, GSHPx-1, Gpx
          Chromosome Location:
          Chr.9: 108339080 - 108340342 on Build GRCm38
          UniGene ID:
          Mm.1090
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s66973

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Gpx1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_008160.6 NP_032186.2 2 789
          GenBank mRNA AK002245.1 - 2 594
          AK010999.1 - 2 594
          AK011019.1 - 2 594
          AK028171.1 - 2 594
          AK150548.1 - 2 595
          AK154833.1 - 2 594
          AK160388.1 - 2 595
          BC086649.1 - 2 577 6819448

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          peroxidase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to reactive oxygen species
          temperature homeostasis
          endothelial cell development
          negative regulation of inflammatory response to antigenic stimulus
          lipid metabolic process
          triglyceride metabolic process
          glutathione metabolic process
          apoptotic process
          response to oxidative stress
          response to lipid hydroperoxide
          sensory perception of sound
          cell proliferation
          intrinsic apoptotic signaling pathway in response to oxidative stress
          response to xenobiotic stimulus
          response to symbiotic bacterium
          response to wounding
          response to toxic substance
          UV protection
          response to selenium ion
          response to gamma radiation
          myotube differentiation
          protein oxidation
          response to hydroperoxide
          regulation of mammary gland epithelial cell proliferation
          regulation of gene expression, epigenetic
          vasodilation
          response to hydrogen peroxide
          hydrogen peroxide catabolic process
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          skeletal muscle tissue regeneration
          regulation of neuron apoptotic process
          blood vessel endothelial cell migration
          fat cell differentiation
          cell redox homeostasis
          skeletal muscle fiber development
          myoblast proliferation
          interaction with symbiont
          positive regulation of protein kinase B signaling
          oxidation-reduction process
          heart contraction
          angiogenesis involved in wound healing
          regulation of proteasomal protein catabolic process
          negative regulation of release of cytochrome c from mitochondria
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
          peroxidase activity
          glutathione peroxidase activity
          protein binding
          selenium binding
          oxidoreductase activity
          SH3 domain binding
          glutathione binding
          phospholipid-hydroperoxide glutathione peroxidase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-5bf9q:80/100.66.78.48:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0