Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grin2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010350.2 | NP_034480.2 | 10 | 2490 | ||
GenBank mRNA | AK147390.1 | - | 9 | 2218 | ||
AK165770.1 | - | 7 | 1345 | |||
BC137849.1 | - | 8 | 1880 | 8860868 | ||
D10694.1 | - | 7 | 1717 |