Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HNRNPC SIRNA Assays ›
Gene Symbol:
HNRNPC
Gene Name:
heterogeneous nuclear ribonucleoprotein C (C1/C2)
Gene Aliases:
C1, C2, HNRNP, HNRPC, SNRPC
Chromosome Location:
Chr.14: 21209136 - 21269479 on Build GRCh38
UniGene ID:
Hs.508848
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s6720
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HNRNPC
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001077442.1
NP_001070910.1
7
907
NM_001077443.1
NP_001070911.1
7
868
NM_004500.3
NP_004491.2
8
894
NM_031314.2
NP_112604.2
8
933
XM_006720125.2
XP_006720188.1
8
1550
XM_011536708.1
XP_011535010.1
9
967
XM_011536709.2
XP_011535011.1
8
1388
XM_011536710.2
XP_011535012.1
8
1589
XM_011536711.2
XP_011535013.1
8
1596
XM_011536712.1
XP_011535014.1
9
928
XM_017021252.1
XP_016876741.1
8
1557
XM_017021253.1
XP_016876742.1
8
1348
GenBank mRNA
AB209527.1
-
8
867
AK126950.1
-
7
3602
AK223517.1
-
8
872
AK225032.1
-
8
857
AK292619.1
-
10
1210
AK297482.1
-
6
733
AK298906.1
-
7
565
AK299923.1
-
7
757
AK302213.1
-
7
831
AK312338.1
-
8
839
AK312382.1
-
7
853
AL833432.1
-
5
470
BC003394.1
-
8
825
3447724
BC007052.1
-
7
853
3686841
BC008364.1
-
8
832
3354131
BC008423.1
-
7
853
4102910
BC066932.1
-
7
813
4375450
BC089438.1
-
7
660
30555817
BC103758.1
-
7
813
6187512
BC108658.1
-
7
758
6501821
BX161480.1
-
7
763
BX247961.1
-
7
624
BX247992.1
-
7
779
M16342.1
-
8
772
M29063.1
-
8
868
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splicing, via spliceosome
osteoblast differentiation
RNA splicing
gene expression
protein sumoylation
negative regulation of telomere maintenance via telomerase
ATP-dependent chromatin remodeling
3'-UTR-mediated mRNA stabilization
nucleotide binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
RNA binding
mRNA 3'-UTR binding
protein binding
poly(U) RNA binding
nucleosomal DNA binding
identical protein binding
poly(A) RNA binding
telomerase RNA binding
N6-methyladenosine-containing RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0