Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gys1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030678.3 | NP_109603.2 | 11, 12 | 1637 | ||
GenBank mRNA | AK050813.1 | - | 9, 10 | 1070 | ||
AK085476.1 | - | 7, 8 | 664 | |||
AK169344.1 | - | 11, 12 | 1633 | |||
AK170404.1 | - | 11, 12 | 1639 | |||
AK170585.1 | - | 11, 12 | 1557 | |||
AK171148.1 | - | 11, 12 | 1592 | |||
AK171426.1 | - | 11, 12 | 1616 | |||
BC019389.1 | - | 11, 12 | 1430 | 3663506 | ||
BC131687.1 | - | 11, 12 | 1484 | 40111722 | ||
BC131688.1 | - | 11, 12 | 1484 | 40111723 | ||
BC152550.1 | - | 11, 12 | 1484 | 40111724 | ||
U53218.1 | - | 11, 12 | 1475 | |||
X94616.1 | - | 11, 12 | 1484 |