Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HNRNPD SIRNA Assays ›
          Gene Symbol:
          HNRNPD
          Gene Name:
          heterogeneous nuclear ribonucleoprotein D
          Gene Aliases:
          AUF1, AUF1A, HNRPD, P37, hnRNPD0
          Chromosome Location:
          Chr.4: 82353314 - 82373996 on Build GRCh38
          UniGene ID:
          Hs.480073
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s6723

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HNRNPD
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001003810.1 NP_001003810.1 4 940
          NM_002138.3 NP_002129.2 5 997
          NM_031369.2 NP_112737.1 4 940
          NM_031370.2 NP_112738.1 5 997
          GenBank mRNA AF039575.1 - 5 655
          AK057836.1 - 5 958
          AK292707.1 - 4 935
          AK300149.1 - 5 829
          AK303552.1 - 3 766
          AL832723.1 - 1 1456
          BC002401.1 - 5 743 2822230
          BC023977.2 - 5 972 2961272
          BC026015.2 - 5 972 2961272
          D55671.1 - 4 416
          D55672.1 - 5 964
          D55673.1 - 4 407
          D55674.1 - 5 943
          X03910.1 - 5 650

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA splicing, via spliceosome
          liver development
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          RNA processing
          RNA catabolic process
          gene expression
          response to fluoxetine
          cerebellum development
          regulation of circadian rhythm
          regulation of mRNA stability
          positive regulation of translation
          positive regulation of transcription, DNA-templated
          mRNA stabilization
          response to calcium ion
          response to electrical stimulus
          3'-UTR-mediated mRNA destabilization
          cellular response to amino acid stimulus
          cellular response to estradiol stimulus
          cellular response to nitric oxide
          circadian regulation of translation
          response to rapamycin
          response to sodium phosphate
          cellular response to putrescine
          hepatocyte dedifferentiation
          nucleotide binding
          AT DNA binding
          chromatin binding
          RNA binding
          protein binding
          transcription factor binding
          mRNA 3'-UTR AU-rich region binding
          telomeric DNA binding
          histone deacetylase binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0