Search
Search

Has2| Gene Symbol: Has2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008216.3 | NP_032242.3 | 2 | 751 | ||
| GenBank mRNA | AK079729.1 | - | 2 | 744 | ||
| BC080281.1 | - | 2 | 706 | 30533251 | ||
| U52524.2 | - | 2 | 705 | |||
| U69695.1 | - | 2 | 725 | |||