Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdgf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008231.4 | NP_032257.3 | 1, 2 | 401 | ||
GenBank mRNA | AF251787.1 | - | 1, 2 | 314 | ||
AK005359.1 | - | 1, 2 | 180 | |||
AK017863.1 | - | 1, 2 | 355 | |||
AK029475.1 | - | 1, 2 | 145 | |||
AK076021.1 | - | 1, 2 | 189 | |||
AK185514.1 | - | 11 | ||||
BC005713.1 | - | 1, 2 | 311 | 2609261 | ||
BC021654.1 | - | 1, 2 | 329 | 4163897 | ||
CJ173614.1 | - | 1, 2 | 401 | |||
D63707.1 | - | 1, 2 | 155 |