Search
Search

Htt| Gene Symbol: Htt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010414.3 | NP_034544.1 | 5, 6 | 723 | ||
| GenBank mRNA | AK042204.1 | - | 5, 6 | 724 | ||
| AK164637.1 | - | 4, 5 | 582 | |||
| L23312.1 | - | 5, 6 | 670 | |||
| L23313.1 | - | 5, 6 | 670 | |||
| L28827.1 | - | 5, 6 | 642 | |||
| U24233.1 | - | 5, 6 | 683 | |||