Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HTT SIRNA Assays ›
          Gene Symbol:
          Htt
          Gene Name:
          huntingtin
          Gene Aliases:
          AI256365, C430023I11Rik, Hd, Hdh, IT15
          Chromosome Location:
          Chr.5: 34761736 - 34912534 on Build GRCm38
          UniGene ID:
          Mm.209071
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s67442

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Htt
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_010414.3 NP_034544.1 21 2828
          GenBank mRNA L23312.1 - 21 2775
          L23313.1 - 21 2775
          L28827.1 - 21 2747
          U24233.1 - 21 2788

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_024357, XM_006251232, XM_008770274, XM_006251230, U18650.

          Gene Ontology Categories:

          Function(s) Process(es)

          urea cycle
          citrulline metabolic process
          establishment of mitotic spindle orientation
          protein import into nucleus
          mitochondrial transport
          ER to Golgi vesicle-mediated transport
          retrograde vesicle-mediated transport, Golgi to ER
          apoptotic process
          mitochondrion organization
          endoplasmic reticulum organization
          Golgi organization
          dopamine receptor signaling pathway
          spermatogenesis
          gastrulation
          central nervous system development
          brain development
          cell aging
          learning or memory
          learning
          grooming behavior
          locomotory behavior
          axo-dendritic transport
          associative learning
          determination of adult lifespan
          visual learning
          anatomical structure morphogenesis
          embryo development
          anterior/posterior pattern specification
          endosomal transport
          lactate biosynthetic process from pyruvate
          quinolinate biosynthetic process
          striatum development
          olfactory lobe development
          neural plate formation
          neurogenesis
          peptide hormone secretion
          insulin secretion
          positive regulation of inositol 1,4,5-trisphosphate-sensitive calcium-release channel activity
          regulation of protein phosphatase type 2A activity
          social behavior
          hormone metabolic process
          negative regulation of apoptotic process
          negative regulation of neuron apoptotic process
          regulation of mitochondrial membrane permeability
          vesicle transport along microtubule
          regulation of synaptic plasticity
          paraxial mesoderm formation
          neuron development
          mRNA transport
          neuron apoptotic process
          response to calcium ion
          regulation of mitochondrial membrane potential
          L-glutamate import
          iron ion homeostasis
          negative regulation of neuron death
          negative regulation of cysteine-type endopeptidase activity
          negative regulation of extrinsic apoptotic signaling pathway
          p53 binding
          receptor binding
          protein binding
          profilin binding
          transcription factor binding
          dynactin binding
          identical protein binding
          ion channel binding
          dynein intermediate chain binding
          beta-tubulin binding
          diazepam binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline