Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hipk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010433.2 | NP_034563.2 | 8 | 2203 | ||
GenBank mRNA | AF077659.1 | - | 8 | 1966 | ||
AF170302.1 | - | 8 | 1953 | |||
AF333792.1 | - | 8 | 1830 |