Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HMGA1 SIRNA Assays ›
          Gene Symbol:
          Hmga1 Hmga1-rs1
          Gene Name:
          high mobility group AT-hook 1
          high mobility group AT-hook I, related sequence 1
          Gene Aliases:
          AL023995, Hmga1a, Hmga1b, Hmgi, Hmgiy, Hmgy
          ENSMUSG00000041059, Hmgi-rs1
          Chromosome Location:
          Chr.17: 27556574 - 27563672 on Build GRCm38
          Chromosome Location:
          Chr.11: 120762794 - 120764392 on Build GRCm38
          UniGene ID:
          Mm.426212
          Mm.426212
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s67596

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hmga1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001025427.3 NP_001020598.1 5 1214
          NM_001039356.2 NP_001034445.1 6 1329
          NM_001166535.1 NP_001160007.1 5 1252
          NM_001166536.1 NP_001160008.1 5 1212
          NM_001166537.1 NP_001160009.1 5 1506
          NM_001166539.1 NP_001160011.1 6 1296
          NM_001166540.1 NP_001160012.1 5 1219
          NM_001166541.1 NP_001160013.1 5 1181
          NM_001166542.1 NP_001160014.1 5 1179
          NM_001166543.1 NP_001160015.1 5 1249
          NM_001166544.1 NP_001160016.1 5 1211
          NM_001166545.1 NP_001160017.1 5 1226
          NM_001166546.1 NP_001160018.1 5 1210
          NM_016660.3 NP_057869.2 5 1539
          GenBank mRNA AK010617.1 - 5 1102
          AK160036.1 - 5 1103
          AK169783.1 - 5 1127
          AK171334.1 - 5 1168
          J04179.1 - 5 1202
          Gene Symbol: Hmga1-rs1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001166476.1 NP_001159948.1 1 1135
          NM_001166477.1 NP_001159949.1 2 1102
          GenBank mRNA BC008125.1 - 2 1087
          BC013455.1 - 1 1145 4219805
          BC093496.1 - 1 1102 3601855

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          endodeoxyribonuclease

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA metabolic process
          base-excision repair
          transcription from RNA polymerase II promoter
          spermatogenesis
          positive regulation of transcription from RNA polymerase II promoter
          nucleic acid phosphodiester bond hydrolysis
          regulation of nucleic acid-templated transcription
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA-(apurinic or apyrimidinic site) lyase activity
          transcription factor binding
          enzyme binding
          ligand-dependent nuclear receptor transcription coactivator activity
          retinoic acid receptor binding
          peroxisome proliferator activated receptor binding
          retinoid X receptor binding
          5'-deoxyribose-5-phosphate lyase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lm92q:80/100.66.76.225:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0