Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HMOX1 SIRNA Assays ›
          Gene Symbol:
          Hmox1
          Gene Name:
          heme oxygenase 1
          Gene Aliases:
          D8Wsu38e, HO-1, HO1, Hemox, Hmox, Hsp32
          Chromosome Location:
          Chr.8: 75093618 - 75100593 on Build GRCm38
          UniGene ID:
          Mm.276389
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s67607

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 404.00
          Your Price
          Online offer: 391.65
          391.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hmox1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_010442.2 NP_034572.1 3 677
          GenBank mRNA AK150934.1 - 3 674
          AK151005.1 - 3 674
          AK151012.1 - 3 666
          AK151627.1 - 3 653
          AK153420.1 - 3 674
          AK153516.1 - 3 674
          AK153566.1 - 3 674
          AK154598.1 - 3 675
          AK159521.1 - 3 674
          AK169227.1 - 3 674
          BC010757.1 - 3 646 4235374
          M33203.1 - 3 619
          X13356.1 - 3 619

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          oxygenase

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          wound healing involved in inflammatory response
          heme oxidation
          cellular iron ion homeostasis
          apoptotic process
          response to oxidative stress
          small GTPase mediated signal transduction
          regulation of blood pressure
          cell death
          negative regulation of cell proliferation
          intrinsic apoptotic signaling pathway in response to DNA damage
          negative regulation of muscle cell apoptotic process
          negative regulation of mast cell cytokine production
          erythrocyte homeostasis
          regulation of transcription from RNA polymerase II promoter in response to iron
          cellular response to heat
          response to nicotine
          intracellular signal transduction
          heme catabolic process
          heme metabolic process
          response to hydrogen peroxide
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of mast cell degranulation
          negative regulation of DNA binding
          negative regulation of sequence-specific DNA binding transcription factor activity
          negative regulation of neuron apoptotic process
          regulation of transcription from RNA polymerase II promoter in response to oxidative stress
          response to estrogen
          positive regulation of angiogenesis
          positive regulation of smooth muscle cell proliferation
          negative regulation of smooth muscle cell proliferation
          regulation of sequence-specific DNA binding transcription factor activity
          protein homooligomerization
          iron ion homeostasis
          oxidation-reduction process
          cellular response to arsenic-containing substance
          cellular response to cadmium ion
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          heme oxygenase (decyclizing) activity
          phospholipase D activity
          signal transducer activity
          oxidoreductase activity
          enzyme binding
          heme binding
          protein homodimerization activity
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-6cc6g:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0