siRNA Details
Gene Symbol: Hoxb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008269.1 | NP_032295.1 | 2 | 1149 | ||
GenBank mRNA | AK011622.1 | - | 2 | 1247 | ||
BC016893.1 | - | 2 | 1149 | 4223365 | ||
X56461.1 | - | 2 | 1662 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
